Assay Details
Gene Symbol: Thap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025780.3 | NP_080056.1 | 2 - 3 | 426 | 123 | |
GenBank mRNA | AK009162.1 | - | 2 - 3 | 405 | 123 | |
AK018577.1 | - | 2 - 3 | 406 | 123 | ||
AK081303.1 | - | 2 - 3 | 427 | 123 | ||
BC027502.1 | - | 2 - 3 | 334 | 1515752 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27469299 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27469296 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27469297 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27469300 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27469302 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P653932 | AK081303.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P653932 | NM_025780.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P653932 | AK081303.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P653932 | NM_025780.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730656 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 739970 | AK004037.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 739970 | AK013196.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 739970 | AK131114.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 739970 | BC022953.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 739970 | NM_025578.4 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 12253 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491135 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573804 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 553125 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 29982 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 660039 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 77786 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 102199 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 115408 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122304 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139814 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P653932 | AK081303.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P653932 | NM_025780.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P653932 | AK081303.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P653932 | NM_025780.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469280 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469286 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469270 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469287 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469302 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469269 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469274 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469292 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469297 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469271 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469285 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469283 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469281 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469300 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469299 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469291 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469290 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469278 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469296 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469273 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469276 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27469295 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07201789 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122304 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139814 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07201800 | - | - |
Whole Mouse Genome G4122A | A_52_P277378 | AK081406.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P653932 | AK081303.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P653932 | NM_025780.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P653932 | AK081303.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P653932 | NM_025780.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P440044 | BC027502.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P440044 | AK081303.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P440044 | AK051198.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P440044 | NM_025780.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P440044 | AK009162.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P118125 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122304 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139814 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08695482 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08695480 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08695488 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27469297 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27469302 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27469299 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27469296 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27469300 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469265 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469290 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469262 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469198 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469202 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469256 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469204 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469187 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469295 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469241 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469230 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469195 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469147 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469297 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27469177 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469266 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469292 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469276 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469286 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469268 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469274 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469265 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469295 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08574456 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08872363 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469262 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469299 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469272 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469297 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469280 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469296 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469291 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469271 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469252 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469287 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469290 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469281 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469255 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469269 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469278 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469283 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469257 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469258 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469285 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27469273 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10372498 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10372499 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10372500 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10372502 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10372501 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1455336_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1421177_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10372497 | - | - |
Mouse Genome 430 2.0 Array | 1421177_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1455336_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1421177_at | - | - |
Mouse Genome 430 2.0 Array | 1455336_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66816_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66816_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1000021899.mm.1 | NM_025780.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000021899.mm.1 | TC1000002902.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000021901.mm.1 | NM_025780.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000021901.mm.1 | TC1000002902.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000021900.mm.1 | NM_025780.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000021900.mm.1 | TC1000002902.mm.1 | MTA TC |
Mouse Expression Set 430 | 1421177_PM_at | - | - |
Mouse Expression Set 430 | 1421177_at | - | - |
Mouse Expression Set 430 | 1455336_at | - | - |
Mouse Expression Set 430 | 1455336_PM_at | - | - |
Murine Genome U74v2 Set | 134709_r_at | - | - |
Murine Genome U74v2 Set | 134709_PM_r_at | - | - |
Mouse Genome 430A 2.0 Array | 1421177_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1421177_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1421177_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1421177_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455336_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455336_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 330730 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6200072 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 104210333 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6200072 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006200072 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1249124 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1252354 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1249124 | - | - |
mouse-6 v1.1 (Illumina) | scl0066816.2_17-S | - | - |
mouse-6 v1.1 (Illumina) | scl37486.4_79-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6200072 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 104210333 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0066816.2_17-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104210333 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6200072 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1249124 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1249124 | - | - |
