This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00469429_g1
See other EIF3D GE Assays ›
Gene Symbol:
Eif3d
Gene Name:
eukaryotic translation initiation factor 3, subunit D
Gene Aliases:
66/67kDa, AA407891, Eif3s7, eIF3p66
Chromosome Location:
Chr.15: 77958998 - 77971088 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00469429_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif3d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018749.2 NP_061219.2 5 - 6 545 86
GenBank mRNA AB012580.1 - 5 - 6 489 86
AK045228.1 - 5 - 6 487 86
AK049767.1 - 5 - 6 484 86
AK145689.1 - 5 - 6 500 86
AK158344.1 - 3 - 4 1491 86
AK168256.1 - 5 - 6 535 86
AK203325.1 - 165 86
BC018598.1 - 5 - 6 483 86
BC089020.1 - 5 - 6 479 30551991 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001824.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif3d

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

formation of cytoplasmic translation initiation complex
translation
translational initiation
translation initiation factor activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500014232.mm.1 NM_018749.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500014232.mm.1 NM_018749.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500014232.mm.1 TC1500001824.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0