This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00469382_g1
See other EIF3G GE Assays ›
Gene Symbol:
Eif3g
Gene Name:
eukaryotic translation initiation factor 3, subunit G
Gene Aliases:
44kDa, D0Jmb4, Eif3s4, TU-189B2, eIF3-p44, p44
Chromosome Location:
Chr.9: 20894349 - 20898590 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00469382_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif3g
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016876.3 NP_058572.2 6 - 7 433 104
GenBank mRNA AF108214.1 - 6 - 7 447 104
AK038249.1 - 6 - 7 443 104
BC008511.1 - 6 - 7 433 3592537 104
U70733.1 - 6 - 7 423 104

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001850.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif3g

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

translation
translational initiation
nucleotide binding
nucleic acid binding
RNA binding
translation initiation factor activity
protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900015496.mm.1 NM_016876.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900015496.mm.1 NM_016876.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900015496.mm.1 TC0900001850.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0