This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00469082_m1
See other EPB4.1L1 GE Assays ›
Gene Symbol:
Epb4.1l1
Gene Name:
erythrocyte protein band 4.1 like 1
Gene Aliases:
4.1N, Epb41l1, NBL1, mKIAA0338
Chromosome Location:
Chr.2: 156420487 - 156543214 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00469082_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Epb4.1l1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001006664.3 NP_001006665.1 16 - 17 2668 114
NM_001291120.1 NP_001278049.1 17 - 18 2635 114
NM_001291121.1 NP_001278050.1 16 - 17 2599 114
NM_001291122.1 NP_001278051.1 15 - 16 2191 114
NM_001291123.1 NP_001278052.1 15 - 16 2188 114
NM_013510.4 NP_038538.1 16 - 17 2586 114
GenBank mRNA AB370262.1 - 15 - 16 2181 114
AF061283.1 - 16 - 17 2400 114
AK122251.1 - 16 - 17 2266 114
AK147272.1 - 15 - 16 2050 114
AK147611.1 - 17 - 18 2637 114
AK154704.1 - 15 - 16 2010 114
BC034751.1 - 16 - 17 2396 4954660 114

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002395.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Epb4.1l1

Gene Ontology Categories:

Function(s) Process(es)

cortical actin cytoskeleton organization
actin binding
receptor binding
structural molecule activity
protein binding
cytoskeletal protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200019289.mm.1 NM_001006664.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200019289.mm.1 NM_013510.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200019289.mm.1 TC0200002395.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm