Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700034075.mm.1 |
NM_001040131.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700034075.mm.1 |
NM_001040131.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700034075.mm.1 |
NM_013507.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700034075.mm.1 |
NM_013507.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700034075.mm.1 |
TC0700004108.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700034076.mm.1 |
NM_001040131.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700034076.mm.1 |
NM_001040131.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700034076.mm.1 |
NM_013507.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700034076.mm.1 |
NM_013507.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700034076.mm.1 |
TC0700004108.mm.1 |
MTA TC |