This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00468700_m1
See other EFS GE Assays ›
Gene Symbol:
Efs
Gene Name:
embryonal Fyn-associated substrate
Gene Aliases:
-
Chromosome Location:
Chr.14: 54916543 - 54926788 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00468700_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Efs
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010112.4 NP_034242.2 1 - 2 1350 91
GenBank mRNA AK030321.1 - 1 - 2 477 91
AK163191.1 - 1 - 2 391 91
BC005438.1 - 1 - 2 419 3487600 91
BC031543.1 - 1 - 2 145 3669513 91
U28728.1 - 1 - 2 374 91
U57686.1 - 1 - 2 74 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002072.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK030321.1,AK163191.1,BC005438.1,BC031543.1,NM_010112.4,U28728.1,U57686.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Efs

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
SH3 domain binding
protein domain specific binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400015666.mm.1 NM_010112.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015666.mm.1 NM_010112.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400015666.mm.1 TC1400002072.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400015667.mm.1 NM_010112.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015667.mm.1 NM_010112.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400015667.mm.1 TC1400002072.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400015669.mm.1 NM_010112.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015669.mm.1 NM_010112.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400015669.mm.1 TC1400002072.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400015668.mm.1 NM_010112.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400015668.mm.1 NM_010112.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400015668.mm.1 TC1400002072.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 921380 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0