Assay Details
Gene Symbol: Dpyd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170778.2 | NP_740748.1 | 16 - 17 | 2162 | 57 | |
GenBank mRNA | BC039699.1 | - | 16 - 17 | 2163 | 4235971 | 57 |
BC042543.1 | - | 12 - 13 | 1627 | 57 | ||
BC044730.2 | - | 4 - 5 | 525 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
