This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00468028_m1
See other CD40 GE Assays ›
Gene Symbol:
Cd40
Gene Name:
CD40 antigen
Gene Aliases:
AI326936, Bp50, GP39, HIGM1, IGM, IMD3, T-BAM, TRAP, Tnfrsf5, p50
Chromosome Location:
Chr.2: 165055604 - 165071654 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00468028_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd40
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_170702.2 NP_733803.2 5 - 6 572 126
GenBank mRNA AJ401390.1 - 5 - 6 506 126

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cd40

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
immune system process
immune response-regulating cell surface receptor signaling pathway
cellular calcium ion homeostasis
inflammatory response
positive regulation of B cell proliferation
positive regulation of interleukin-12 production
B cell activation
positive regulation of tyrosine phosphorylation of Stat1 protein
defense response to protozoan
positive regulation of MAP kinase activity
protein kinase B signaling
positive regulation of GTPase activity
positive regulation of transcription from RNA polymerase II promoter
positive regulation of isotype switching to IgG isotypes
regulation of immune response
regulation of immunoglobulin secretion
positive regulation of NF-kappaB transcription factor activity
defense response to virus
positive regulation of protein kinase C signaling
positive regulation of endothelial cell apoptotic process
antigen binding
protein binding
enzyme binding
ubiquitin protein ligase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0