This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00467386_m1
See other ZFP410 GE Assays ›
Gene Symbol:
Zfp410
Gene Name:
zinc finger protein 410
Gene Aliases:
Apa1, D12Ertd748e, Znf410
Chromosome Location:
Chr.12: 84315928 - 84344439 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00467386_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zfp410
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252582.1 NP_001239511.1 5 - 6 1078 65
NM_001252583.1 NP_001239512.1 5 - 6 1043 65
NM_144833.3 NP_659082.1 5 - 6 1043 65
NR_045546.1 5 - 6 1011 65
NR_045615.1 3 - 4 727 65
GenBank mRNA AF295806.1 - 5 - 6 805 65
AK033090.1 - 5 - 6 820 65
AK042224.1 - 5 - 6 762 65
AK051939.1 - 3 - 4 443 65
AK053001.1 - 5 - 6 768 65
AK083003.1 - 5 - 6 786 65
AK132345.1 - 5 - 6 781 65
AK139797.1 - 4 - 5 597 65
BC021528.1 - 5 - 6 798 5353673 65
BC052080.1 - 5 - 6 775 6403411 65
BC056957.1 - 5 - 6 740 65

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Zfp410

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
nucleic acid binding
DNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0