This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00465967_m1
See other S100A14 GE Assays ›
Gene Symbol:
S100a14
Gene Name:
S100 calcium binding protein A14
Gene Aliases:
1110013O05Rik, S100a15, S114
Chromosome Location:
Chr.3: 90526849 - 90528837 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00465967_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100a14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163525.2 NP_001156997.1 2 - 3 188 72
NM_025393.4 NP_079669.2 2 - 3 171 72
GenBank mRNA AA726094.1 - 2 - 3 181 72
AK003669.1 - 2 - 3 151 72
AK019075.1 - 2 - 3 142 72
BC025607.1 - 1 - 2 170 5151924 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000801.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK003669.1,AK019075.1,BC025607.1,NM_001163525.2,NM_025393.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene S100a14

Panther Classification:

Molecular Function -

calmodulin-related

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
toll-like receptor 4 signaling pathway
positive regulation of granulocyte chemotaxis
positive regulation of monocyte chemotaxis
calcium ion binding
chemokine receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300006253.mm.1 NM_001163525.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300006253.mm.1 NM_001163525.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300006253.mm.1 NM_025393.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300006253.mm.1 NM_025393.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300006253.mm.1 TC0300000801.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0