This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00464314_m1
See other ARHGAP15 GE Assays ›
Gene Symbol:
Arhgap15
Gene Name:
Rho GTPase activating protein 15
Gene Aliases:
5830480G12Rik
Chromosome Location:
Chr.2: 43748783 - 44395953 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00464314_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153820.3 NP_722542.2 8 - 9 783 91
GenBank mRNA AK040796.1 - 9 - 10 853 91
AK170537.1 - 8 - 9 781 91
BC024887.1 - 8 - 9 737 3156334 91
BC034881.1 - 8 - 9 756 3468639 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000741.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgap15

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
regulation of cell shape
positive regulation of GTPase activity
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200006407.mm.1 NM_153820.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200006407.mm.1 NM_153820.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200006407.mm.1 TC0200000741.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200006405.mm.1 NM_153820.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200006405.mm.1 NM_153820.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200006405.mm.1 TC0200000741.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200006406.mm.1 NM_153820.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200006406.mm.1 NM_153820.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200006406.mm.1 TC0200000741.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0