This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00464288_m1
See other DNTTIP2 GE Assays ›
Gene Symbol:
Dnttip2
Gene Name:
deoxynucleotidyltransferase, terminal, interacting protein 2
Gene Aliases:
4930588M11Rik, AA408582, AU014960, HSU15552
Chromosome Location:
Chr.3: 122274414 - 122285271 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00464288_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dnttip2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153806.1 NP_722501.1 1 - 2 66 73
GenBank mRNA AK016368.1 - 1 - 2 59 73
AK133904.1 - 1 - 2 85 73
AK135957.1 - 1 - 2 84 73
AK168536.1 - 1 - 2 85 73
BC028305.1 - 1 - 2 66 2609541 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001245.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dnttip2

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300010162.mm.1 NM_153806.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300010162.mm.1 NM_153806.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300010162.mm.1 TC0300001245.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300010161.mm.1 NM_153806.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300010161.mm.1 NM_153806.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300010161.mm.1 TC0300001245.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0