This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00463845_m1
See other CNOT10 GE Assays ›
Gene Symbol:
Cnot10
Gene Name:
CCR4-NOT transcription complex, subunit 10
Gene Aliases:
2600001P13Rik
Chromosome Location:
Chr.9: 114585874 - 114640216 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00463845_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cnot10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153585.5 NP_705813.2 8 - 9 1122 65
GenBank mRNA AK029103.1 - 8 - 9 997 65
AK149944.1 - 8 - 9 955 65
AK157824.1 - 8 - 9 1021 65
AK171701.1 - 8 - 9 1102 65
BC025554.1 - 8 - 9 983 5321869 65
BC036178.1 - 5 - 6 578 65
BC036179.1 - 8 - 9 1094 5059723 65
BC039183.1 - 8 - 9 1096 2631793 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900003172.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cnot10

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of translation
biological_process
gene silencing by RNA
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900026867.mm.1 NM_153585.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900026867.mm.1 NM_153585.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900026867.mm.1 TC0900003172.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900026868.mm.1 NM_153585.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900026868.mm.1 NM_153585.5 Refseq
Affymetrix Mouse Clariom S Array PSR0900026868.mm.1 TC0900003172.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0