This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00463772_g1
See other NOC4L GE Assays ›
Gene Symbol:
Noc4l
Gene Name:
nucleolar complex associated 4 homolog (S. cerevisiae)
Gene Aliases:
AI326906
Chromosome Location:
Chr.5: 110648419 - 110653382 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00463772_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Noc4l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153570.2 NP_705798.2 13 - 14 1363 76
GenBank mRNA AK052601.1 - 13 - 14 1363 76
AK090024.1 - 13 - 14 1375 76
AK146006.1 - 13 - 14 1359 76
AK172310.1 - 13 - 14 1342 76
BC024616.1 - 13 - 14 1353 4218310 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002993.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Noc4l

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
ribosome biogenesis
poly(A) RNA binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23981086 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23981083 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23981081 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23981085 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23981084 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23981082 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P346453 AK146006.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P346453 AK172310.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P346453 BC024616.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P346453 AK088355.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P346453 AK052601.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P346453 AK146006.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P346453 AK172310.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P346453 BC024616.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P346453 AK088355.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P346453 AK052601.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 423515 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753574 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806686 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 60690 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174140 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P346453 AK146006.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P346453 AK172310.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P346453 BC024616.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P346453 AK088355.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P346453 AK052601.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P346453 AK146006.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P346453 AK172310.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P346453 BC024616.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P346453 AK088355.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P346453 AK052601.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981073 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981043 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981054 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981086 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981057 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981075 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981065 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981070 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981085 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981060 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981048 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981051 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981052 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981042 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981064 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981050 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981056 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981044 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981049 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981071 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981079 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981083 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981062 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981058 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981061 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981081 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981046 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23981068 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174140 - -
Whole Mouse Genome G4122A A_51_P346453 AK146006.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P346453 AK172310.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P346453 BC024616.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P346453 AK088355.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P346453 AK052601.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P346453 AK146006.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P346453 AK172310.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P346453 BC024616.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P346453 AK088355.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P346453 AK052601.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105620 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123662 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174140 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04940610 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04987370 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23981082 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23981081 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23981084 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23981086 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23981085 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23981083 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981050 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981060 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981048 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981038 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981068 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981043 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981057 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981040 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981035 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981042 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981031 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981062 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981056 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981052 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981070 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981064 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981071 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981037 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981086 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981051 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981083 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981058 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981079 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981073 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981033 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981054 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981032 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981029 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981065 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981075 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981046 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981085 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23981081 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04940633 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23981056 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04940630 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23981081 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23981058 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04940607 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04940610 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23981043 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23981038 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04940627 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23981042 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04940616 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23981037 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23981065 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23981070 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04940604 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23981040 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04940631 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23981083 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04940594 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04940609 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P23981073 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04940634 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04940636 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04940625 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P04940596 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10532395 - -
Affymetrix Mouse Gene 1.0 ST Array 10532398 - -
Affymetrix Mouse Gene 1.0 ST Array 10532406 - -
Affymetrix Mouse Gene 1.0 ST Array 10532397 - -
Affymetrix Mouse Gene 1.0 ST Array 10532403 - -
Affymetrix Mouse Gene 1.0 ST Array 10532404 - -
Affymetrix Mouse Gene 1.0 ST Array 10532399 - -
Affymetrix Mouse Gene 1.0 ST Array 10532401 - -
Affymetrix Mouse Gene 1.0 ST Array 10532393 - -
Affymetrix Mouse Gene 1.0 ST Array 10532405 - -
Affymetrix Mouse Gene 1.0 ST Array 10532400 - -
Affymetrix Mouse Gene 1.0 ST Array 10532394 - -
Affymetrix Mouse Gene 1.0 ST Array 10532392 - -
Affymetrix Mouse Gene 1.0 ST Array 10532391 - -
Affymetrix Mouse Gene 1.0 ST Array 10532396 - -
Affymetrix Mouse Gene 1.0 ST Array 10532402 - -
Affymetrix HT MG-430 PM Array Plate 1423827_PM_s_at - -
Affymetrix HT MG-430 PM Array Plate 1423826_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1438095_PM_x_at - -
Affymetrix HT MG-430 PM Array Plate 1443794_PM_x_at - -
Affymetrix Mouse Gene 1.1 ST Array 10532390 - -
Mouse Genome 430 2.0 Array 1443794_PM_x_at - -
Mouse Genome 430 2.0 Array 1438095_x_at - -
Mouse Genome 430 2.0 Array 1423826_PM_at - -
Mouse Genome 430 2.0 Array 1423827_PM_s_at - -
Mouse Genome 430 2.0 Array 1438095_PM_x_at - -
Mouse Genome 430 2.0 Array 1423827_s_at - -
Mouse Genome 430 2.0 Array 1443794_x_at - -
Mouse Genome 430 2.0 Array 1423826_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 100608_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 100608_at - -
Affymetrix Mouse Clariom S Array PSR0500026473.mm.1 NM_153570.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026473.mm.1 NM_153570.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500026473.mm.1 TC0500002993.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500026476.mm.1 NM_153570.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026476.mm.1 NM_153570.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500026476.mm.1 TC0500002993.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500026472.mm.1 NM_153570.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026472.mm.1 NM_153570.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500026472.mm.1 TC0500002993.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500026474.mm.1 NM_153570.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026474.mm.1 NM_153570.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500026474.mm.1 TC0500002993.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500026475.mm.1 NM_153570.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026475.mm.1 NM_153570.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500026475.mm.1 TC0500002993.mm.1 MTA TC
Mouse Expression Set 430 1438095_x_at - -
Mouse Expression Set 430 1438095_PM_x_at - -
Mouse Expression Set 430 1443794_x_at - -
Mouse Expression Set 430 1443794_PM_x_at - -
Murine Genome U74v2 Set 97414_PM_at - -
Murine Genome U74v2 Set 161241_PM_at - -
Murine Genome U74v2 Set 161241_at - -
Murine Genome U74v2 Set 97414_at - -
Mouse Genome 430A 2.0 Array 1438095_PM_x_at - -
Mouse Genome 430A 2.0 Array 1438095_x_at - -
Affymetrix HT Mouse Genome 430A Array 1423826_at - -
Affymetrix HT Mouse Genome 430A Array 1423827_PM_s_at - -
Affymetrix HT Mouse Genome 430A Array 1438095_PM_x_at - -
Affymetrix HT Mouse Genome 430A Array 1423827_s_at - -
Affymetrix HT Mouse Genome 430A Array 1438095_x_at - -
Affymetrix HT Mouse Genome 430A Array 1423826_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1443794_x_at - -
Affymetrix HT Mouse Genome 430B Array 1443794_PM_x_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 70541 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000070541 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2600627 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2600627 - -
mouse-6 v1.1 (Illumina) scl26224.14.1_3-S - -
Illumina Sentrix mouse-6.v1 bead array 70541 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl26224.14.1_3-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 70541 - -
MouseRef-8 v2 Expression BeadChip ILMN_2957862 - -
MouseRef-8 v2 Expression BeadChip ILMN_2600627 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2600627 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2957862 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl26224.14.1_3-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0