This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00462092_g1
See other CTDSP1 GE Assays ›
Gene Symbol:
Ctdsp1
Gene Name:
CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase 1
Gene Aliases:
GIP, NLIIF, Nif3, SCP1
Chromosome Location:
Chr.1: 74391609 - 74397285 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00462092_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ctdsp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153088.2 NP_694728.1 5 - 6 763 77
GenBank mRNA AK089478.1 - 5 - 6 671 77
AK167390.1 - 5 - 6 521 77
AY028804.1 - 5 - 6 788 77
BC049184.1 - 5 - 6 623 6439402 77
BC058085.1 - 4 - 5 950 77
BC065158.1 - 5 - 6 498 5717102 77
BC079638.1 - 5 - 6 498 5717102 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000633.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ctdsp1

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein phosphorylation
regulation of transcription from RNA polymerase II promoter
protein dephosphorylation
negative regulation of neuron differentiation
negative regulation of neurogenesis
negative regulation of G1/S transition of mitotic cell cycle
phosphoprotein phosphatase activity
protein binding
CTD phosphatase activity
hydrolase activity
phosphatase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100005121.mm.1 NM_153088.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100005121.mm.1 NM_153088.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100005121.mm.1 TC0100000633.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0