This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00461436_m1
See other SLC5A11 GE Assays ›
Gene Symbol:
Slc5a11
Gene Name:
solute carrier family 5 (sodium/glucose cotransporter), member 11
Gene Aliases:
2010013B02Rik, Kst1, RKST2, SMIT2, Slc5a10
Chromosome Location:
Chr.7: 123214758 - 123274390 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00461436_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc5a11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146198.2 NP_666310.1 7 - 8 746 66
GenBank mRNA AF518726.1 - 8 - 9 839 66
AK008219.1 - 7 - 8 800 66
BC031742.1 - 7 - 8 747 4221537 66

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc5a11

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
sodium ion transport
apoptotic process
carbohydrate transport
transmembrane transport
transporter activity
symporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0