This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00461064_g1
See other ALG12 GE Assays ›
Gene Symbol:
Alg12
Gene Name:
asparagine-linked glycosylation 12 (alpha-1,6-mannosyltransferase)
Gene Aliases:
ECM39
Chromosome Location:
Chr.15: 88805243 - 88819318 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00461064_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Alg12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001142357.1 NP_001135829.1 9 - 10 1314 67
NM_145477.2 NP_663452.1 7 - 8 1011 67
GenBank mRNA AJ429133.1 - 9 - 10 1274 67
AK142322.1 - 9 - 10 1315 67
BC021379.1 - 7 - 8 984 5044319 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002031.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Alg12

Panther Classification:

Molecular Function -

glycosyltransferase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein N-linked glycosylation
dolichol-linked oligosaccharide biosynthetic process
mannosylation
mannosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
dol-P-Man:Man(7)GlcNAc(2)-PP-Dol alpha-1,6-mannosyltransferase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30421419 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30421416 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30421420 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30421417 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30421418 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115781 BC021379.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115781 AK142322.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115781 NM_145477.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115781 NM_001142357.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 213461 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358047 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 533355 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 97018 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 129941 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838083 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735069 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 402080 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 13330 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 120543 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140953 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166870 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141393 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115781 BC021379.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115781 AK142322.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115781 NM_145477.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115781 NM_001142357.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P245248 BC021379.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P245248 AK142322.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P245248 NM_145477.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P245248 NM_001142357.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P549125 BC021379.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P549125 AJ429133.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P549125 AK142322.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P549125 NM_145477.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P549125 NM_001142357.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30421393 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30421403 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30421414 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30421411 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30421409 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30421404 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30421417 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30421407 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30421395 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30421418 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30421401 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30421400 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30421398 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30421415 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30421419 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30421413 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30421397 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140953 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08453683 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166870 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141393 - -
Whole Mouse Genome G4122A A_52_P549125 BC021379.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P549125 AJ429133.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P549125 AK142322.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P549125 NM_145477.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P549125 NM_001142357.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P245248 BC021379.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P245248 AK142322.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P245248 NM_145477.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P245248 NM_001142357.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131416 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111169 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140953 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141393 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166870 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12386570 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12386565 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12386569 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12386571 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30421418 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30421416 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30421419 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30421417 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30421420 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421358 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421378 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421338 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421330 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421375 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421373 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421417 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421409 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421401 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421398 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421346 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421348 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421397 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421384 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421411 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421324 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421335 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421318 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421418 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421344 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421323 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421380 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421326 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421390 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421342 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421341 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421414 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421331 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421345 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421367 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421382 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421400 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421356 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421334 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421403 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421327 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421351 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421395 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421357 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421360 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421383 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421376 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421371 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421321 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30421370 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12196235 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421405 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421374 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421384 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421398 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421409 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421375 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421386 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421418 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421378 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421417 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421414 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421415 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12077599 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12444109 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421371 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421376 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421397 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421413 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421403 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11925917 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11849830 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11972299 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421401 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421390 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11925922 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12423743 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421400 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30421383 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11978373 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11995787 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10431303 - -
Affymetrix Mouse Gene 1.0 ST Array 10431301 - -
Affymetrix Mouse Gene 1.0 ST Array 10431304 - -
Affymetrix Mouse Gene 1.0 ST Array 10431310 - -
Affymetrix Mouse Gene 1.0 ST Array 10431302 - -
Affymetrix Mouse Gene 1.0 ST Array 10431308 - -
Affymetrix Mouse Gene 1.0 ST Array 10431309 - -
Affymetrix Mouse Gene 1.0 ST Array 10431307 - -
Affymetrix Mouse Gene 1.0 ST Array 10431305 - -
Affymetrix Mouse Gene 1.0 ST Array 10431306 - -
Affymetrix HT MG-430 PM Array Plate 1424818_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1427558_PM_s_at - -
Affymetrix HT MG-430 PM Array Plate 1427557_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10431300 - -
Mouse Genome 430 2.0 Array 1424818_at - -
Mouse Genome 430 2.0 Array 1427557_at - -
Mouse Genome 430 2.0 Array 1427558_PM_s_at - -
Mouse Genome 430 2.0 Array 1427557_PM_at - -
Mouse Genome 430 2.0 Array 1427558_s_at - -
Mouse Genome 430 2.0 Array 1424818_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 223774_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 223774_at - -
Affymetrix Mouse Clariom S Array PSR1500016073.mm.1 NM_001142357.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016073.mm.1 NM_001142357.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016073.mm.1 NM_145477.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016073.mm.1 NM_145477.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500016073.mm.1 TC1500002031.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500016075.mm.1 NM_001142357.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016075.mm.1 NM_001142357.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016075.mm.1 TC1500002031.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500016078.mm.1 NM_001142357.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016078.mm.1 NM_001142357.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016078.mm.1 NM_145477.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016078.mm.1 NM_145477.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500016078.mm.1 TC1500002031.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500016083.mm.1 NM_001142357.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016083.mm.1 NM_001142357.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016083.mm.1 NM_145477.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016083.mm.1 NM_145477.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500016083.mm.1 TC1500002031.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500016084.mm.1 NM_001142357.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016084.mm.1 NM_001142357.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016084.mm.1 NM_145477.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016084.mm.1 NM_145477.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500016084.mm.1 TC1500002031.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500016072.mm.1 NM_001142357.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016072.mm.1 NM_001142357.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016072.mm.1 NM_145477.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016072.mm.1 NM_145477.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500016072.mm.1 TC1500002031.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500016076.mm.1 NM_145477.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016076.mm.1 NM_145477.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500016076.mm.1 TC1500002031.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500016081.mm.1 NM_001142357.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016081.mm.1 NM_001142357.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016081.mm.1 NM_145477.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016081.mm.1 NM_145477.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500016081.mm.1 TC1500002031.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500016080.mm.1 NM_001142357.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016080.mm.1 NM_001142357.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016080.mm.1 NM_145477.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016080.mm.1 NM_145477.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500016080.mm.1 TC1500002031.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500016071.mm.1 NM_001142357.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016071.mm.1 NM_001142357.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016071.mm.1 NM_145477.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016071.mm.1 NM_145477.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500016071.mm.1 TC1500002031.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500016074.mm.1 NM_001142357.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016074.mm.1 NM_001142357.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016074.mm.1 TC1500002031.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500016077.mm.1 NM_001142357.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016077.mm.1 NM_001142357.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016077.mm.1 NM_145477.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016077.mm.1 NM_145477.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500016077.mm.1 TC1500002031.mm.1 MTA TC
Mouse Expression Set 430 1427558_s_at - -
Mouse Expression Set 430 1424818_PM_at - -
Mouse Expression Set 430 1427558_PM_s_at - -
Mouse Expression Set 430 1427557_PM_at - -
Mouse Expression Set 430 1424818_at - -
Mouse Expression Set 430 1427557_at - -
Mouse Genome 430A 2.0 Array 1427558_s_at - -
Mouse Genome 430A 2.0 Array 1427557_PM_at - -
Mouse Genome 430A 2.0 Array 1424818_at - -
Mouse Genome 430A 2.0 Array 1424818_PM_at - -
Mouse Genome 430A 2.0 Array 1427557_at - -
Mouse Genome 430A 2.0 Array 1427558_PM_s_at - -
Affymetrix HT Mouse Genome 430A Array 1427557_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1427557_at - -
Affymetrix HT Mouse Genome 430A Array 1424818_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1427558_s_at - -
Affymetrix HT Mouse Genome 430A Array 1427558_PM_s_at - -
Affymetrix HT Mouse Genome 430A Array 1424818_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 352119 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 1400692 - -
Illumina mouse-6 v1.0 expression beadchip 1400692 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001400692 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2680684 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2680684 - -
mouse-6 v1.1 (Illumina) scl46865.11.1_0-S - -
Illumina Sentrix mouse-6.v1 bead array 1400692 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl46865.11.1_0-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1400692 - -
MouseRef-8 v2 Expression BeadChip ILMN_2680686 - -
MouseRef-8 v2 Expression BeadChip ILMN_2869182 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2869182 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2680686 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0