This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00460997_m1
See other EIF2B1 GE Assays ›
Gene Symbol:
Eif2b1
Gene Name:
eukaryotic translation initiation factor 2B, subunit 1 (alpha)
Gene Aliases:
26kDa, D5Ertd406e, EIF2B, EIF2BA
Chromosome Location:
Chr.5: 124570214 - 124579057 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00460997_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif2b1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145371.3 NP_663346.1 4 - 5 411 83
GenBank mRNA AK088066.1 - 4 - 5 410 83
AK133695.1 - 4 - 5 412 83
BC003426.1 - 4 - 5 411 2615801 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003261.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Endogenous Controls Mus musculus Mouse Endogenous Controls Inventoried Mouse_Endogenous_Controls Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif2b1

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

translation
translational initiation
regulation of translation
regulation of translational initiation
response to heat
response to glucose
oligodendrocyte development
negative regulation of translational initiation in response to stress
response to peptide hormone
positive regulation of GTPase activity
cellular metabolic process
cellular response to stimulus
translation initiation factor activity
guanyl-nucleotide exchange factor activity
protein binding
GTP binding
GDP binding
enzyme regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0