This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00460854_m1
See other EIF3L GE Assays ›
Gene Symbol:
Eif3l
Gene Name:
eukaryotic translation initiation factor 3, subunit L
Gene Aliases:
0610011H21Rik, D15N1e, Eif3eip, Eif3ip, Eif3s6ip, HSP-66Y, PAF67
Chromosome Location:
Chr.15: 79075223 - 79094400 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00460854_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif3l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145139.2 NP_660121.2 3 - 4 329 78
GenBank mRNA AJ310346.1 - 3 - 4 339 78
AK032936.1 - 3 - 4 317 78
AK075584.1 - 3 - 4 335 78
AK150895.1 - 3 - 4 324 78
AK151175.1 - 3 - 4 324 78
AK167640.1 - 3 - 4 324 78
AK168009.1 - 3 - 4 327 78
AK168295.1 - 3 - 4 326 78
BC024463.1 - 3 - 4 329 4975621 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000665.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif3l

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translation
translational initiation
translation initiation factor activity
protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500004944.mm.1 NM_145139.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004944.mm.1 NM_145139.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500004944.mm.1 TC1500000665.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500004943.mm.1 NM_145139.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004943.mm.1 NM_145139.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500004943.mm.1 TC1500000665.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0