This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00460598_m1
See other SUSD4 GE Assays ›
Gene Symbol:
Susd4
Gene Name:
sushi domain containing 4
Gene Aliases:
AI848994, E430021N18Rik, N28096
Chromosome Location:
Chr.1: 182763865 - 182896596 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00460598_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Susd4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144796.4 NP_659045.2 5 - 6 1054 96
GenBank mRNA AK035779.1 - 5 - 6 1054 96
AK088629.1 - 5 - 6 967 96
BC021842.1 - 5 - 6 1070 5135798 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001752.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Susd4

Gene Ontology Categories:

Function(s) Process(es)

regulation of complement activation
negative regulation of complement activation, alternative pathway
negative regulation of complement activation, classical pathway
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100014629.mm.1 NM_144796.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100014629.mm.1 NM_144796.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100014629.mm.1 TC0100001752.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0