Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800013325.mm.1 |
NM_133238.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013325.mm.1 |
NM_133238.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013325.mm.1 |
TC0800001619.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800013324.mm.1 |
TC0800001619.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800013326.mm.1 |
NM_133238.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013326.mm.1 |
NM_133238.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013326.mm.1 |
TC0800001619.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800013322.mm.1 |
NM_133238.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013322.mm.1 |
NM_133238.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013322.mm.1 |
TC0800001619.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800013323.mm.1 |
NM_133238.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013323.mm.1 |
NM_133238.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013323.mm.1 |
TC0800001619.mm.1 |
MTA TC |