Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10576830 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10576833 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10576831 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10576832 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10576834 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1421562_PM_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10576829 |
- |
- |
Mouse Genome 430 2.0 Array |
1421562_at |
- |
- |
Mouse Genome 430 2.0 Array |
1421562_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
170776_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
170776_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0800013380.mm.1 |
NM_130903.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013380.mm.1 |
NM_130903.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013380.mm.1 |
TC0800001625.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800013381.mm.1 |
NM_130903.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013381.mm.1 |
NM_130903.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013381.mm.1 |
TC0800001625.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800013378.mm.1 |
NM_130903.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013378.mm.1 |
NM_130903.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013378.mm.1 |
TC0800001625.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800013382.mm.1 |
NM_130903.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013382.mm.1 |
NM_130903.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013382.mm.1 |
TC0800001625.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800013376.mm.1 |
NM_130903.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013376.mm.1 |
NM_130903.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013376.mm.1 |
TC0800001625.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800013375.mm.1 |
NM_130903.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013375.mm.1 |
NM_130903.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013375.mm.1 |
TC0800001625.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800013374.mm.1 |
NM_130903.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013374.mm.1 |
NM_130903.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013374.mm.1 |
TC0800001625.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800013377.mm.1 |
NM_130903.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013377.mm.1 |
NM_130903.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013377.mm.1 |
TC0800001625.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800013379.mm.1 |
NM_130903.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013379.mm.1 |
NM_130903.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013379.mm.1 |
TC0800001625.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1421562_at |
- |
- |
Mouse Expression Set 430 |
1421562_PM_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1421562_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1421562_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1421562_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1421562_PM_at |
- |
- |