This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00459309_m1
See other RIN2 GE Assays ›
Gene Symbol:
Rin2
Gene Name:
Ras and Rab interactor 2
Gene Aliases:
2010003K16Rik, 4632403N06Rik, AW821980, RASSF4
Chromosome Location:
Chr.2: 145771727 - 145887616 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00459309_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rin2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028724.4 NP_083000.5 11 - 12 2480 64
GenBank mRNA AK014548.1 - 3 - 4 952 64
AK040763.1 - 3 - 4 952 64
AK150680.1 - 9 - 10 2288 64
AK152893.1 - 9 - 10 2288 64
AK154421.1 - 3 - 4 896 64
AK163047.1 - 3 - 4 1467 64
AK171064.1 - 3 - 4 867 64
AK198881.1 - 2 - 3 267 64
BC005529.1 - 3 - 4 913 3495681 64
BC040390.1 - 11 - 12 2474 3156682 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002166.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rin2

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
signal transduction
biological_process
molecular_function
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200017116.mm.1 NM_028724.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017116.mm.1 NM_028724.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200017116.mm.1 TC0200002166.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0