This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00459209_m1
See other XRCC4 GE Assays ›
Gene Symbol:
Xrcc4
Gene Name:
X-ray repair complementing defective repair in Chinese hamster cells 4
Gene Aliases:
2310057B22Rik, AW413319, AW545101
Chromosome Location:
Chr.13: 89848904 - 90089608 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00459209_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Xrcc4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028012.4 NP_082288.1 3 - 4 435 126
GenBank mRNA AK009951.1 - 3 - 4 352 126
AK053612.1 - 351 126

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Xrcc4

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
pro-B cell differentiation
DNA ligation
DNA repair
double-strand break repair
double-strand break repair via nonhomologous end joining
DNA recombination
cellular response to DNA damage stimulus
central nervous system development
response to X-ray
response to ionizing radiation
response to gamma radiation
T cell differentiation in thymus
V(D)J recombination
immunoglobulin V(D)J recombination
negative regulation of neuron apoptotic process
isotype switching
positive regulation of fibroblast proliferation
positive regulation of neurogenesis
DNA ligation involved in DNA repair
positive regulation of ligase activity
DNA binding
protein C-terminus binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0