This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00459205_g1
See other CIC GE Assays ›
Gene Symbol:
Cic
Gene Name:
capicua homolog (Drosophila)
Gene Aliases:
1200010B10Rik, mKIAA0306
Chromosome Location:
Chr.7: 25267657 - 25294159 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00459205_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cic
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AB093224.1 - 12 - 13 3489 70
AF363690.1 - 17 - 18 4225 70
AK004685.1 - 8 - 9 1618 70
AK082152.1 - 7 - 8 1379 70
AK147276.1 - 17 - 18 4852 70
AK147535.1 - 18 - 19 6967 70
BC040463.1 - 8 - 9 1916 70
BC058665.1 - 14 - 15 3737 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0