This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00459192_g1
See other CIC GE Assays ›
Gene Symbol:
Cic
Gene Name:
capicua homolog (Drosophila)
Gene Aliases:
1200010B10Rik, mKIAA0306
Chromosome Location:
Chr.7: 25267657 - 25294159 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00459192_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cic
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF363690.1 - 2 - 3 252 54
AK147276.1 - 2 - 3 885 54
AK147535.1 - 3 - 4 3003 54

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 299826 - -
ABI Mouse Genome Survey Microarray 764650 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0