This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00458977_m1
See other RGS19 GE Assays ›
Gene Symbol:
Rgs19
Gene Name:
regulator of G-protein signaling 19
Gene Aliases:
2610042F04Rik, AI324841, AW547781, GAIP
Chromosome Location:
Chr.2: 181688419 - 181693977 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00458977_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rgs19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291209.1 NP_001278138.1 1 - 2 105 57
NM_026446.4 NP_080722.1 1 - 2 105 57
GenBank mRNA AK011740.2 - 1 - 2 54 57
AK019401.1 - 1 - 2 78 57
AK042531.1 - 1 - 2 53 57
BC003838.1 - 1 - 2 37 3490927 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200005454.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK019401.1,AK042531.1,BC003838.1,NM_001291209.1,NM_026446.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rgs19

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of signal transduction
termination of G-protein coupled receptor signaling pathway
positive regulation of GTPase activity
G-protein alpha-subunit binding
GTPase activator activity
protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200046616.mm.1 NM_026446.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200046616.mm.1 TC0200005454.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200046620.mm.1 NM_026446.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200046620.mm.1 TC0200005454.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 620272 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0