Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300009895.mm.1 |
NM_019429.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300009895.mm.1 |
NM_019429.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300009895.mm.1 |
TC1300001573.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300009899.mm.1 |
NM_019429.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300009899.mm.1 |
NM_019429.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300009899.mm.1 |
TC1300001573.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300009896.mm.1 |
NM_019429.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300009896.mm.1 |
NM_019429.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300009896.mm.1 |
TC1300001573.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300009897.mm.1 |
NM_019429.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300009897.mm.1 |
NM_019429.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300009897.mm.1 |
TC1300001573.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300009893.mm.1 |
NM_019429.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300009893.mm.1 |
NM_019429.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300009893.mm.1 |
TC1300001573.mm.1 |
MTA TC |