This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00456288_gH
See other STMN3 GE Assays ›
Gene Symbol:
Stmn3
Gene Name:
stathmin-like 3
Gene Aliases:
9330161A03Rik, AI604839, R75323, Sclip
Chromosome Location:
Chr.2: 181306459 - 181314500 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00456288_gH
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stmn3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009133.3 NP_033159.1 4 - 5 768 87
GenBank mRNA AB007912.1 - 4 - 5 556 87
AF069708.1 - 4 - 5 537 87
AK002957.1 - 4 - 5 526 87
AK003203.1 - 4 - 5 526 87
AK032737.1 - 4 - 5 769 87
BC057017.1 - 4 - 5 544 6818794 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200005442.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stmn3

Gene Ontology Categories:

Function(s) Process(es)

microtubule depolymerization
regulation of microtubule polymerization or depolymerization
cytoplasmic microtubule organization
neuron projection development
negative regulation of Rac protein signal transduction
regulation of GTPase activity
regulation of cytoskeleton organization
protein binding
protein C-terminus binding
tubulin binding
protein domain specific binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140690 AK149129.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140690 BC057017.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140690 AB007912.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140690 AF069708.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140690 AK002957.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140690 AK149129.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140690 BC057017.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140690 AB007912.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140690 AF069708.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P140690 AK002957.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 828835 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 699776 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 824933 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 525910 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 713919 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 125413 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 136288 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153561 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140690 AK149129.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140690 BC057017.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140690 AB007912.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140690 AF069708.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140690 AK002957.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140690 AK149129.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140690 BC057017.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140690 AB007912.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140690 AF069708.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P140690 AK002957.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909823 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909866 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909869 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909852 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909858 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909838 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909856 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909853 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909864 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909828 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909826 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909861 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909837 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909848 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909865 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909862 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909840 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909830 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909832 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909849 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909842 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909855 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909835 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909845 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909834 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909867 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909822 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909833 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909839 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909846 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909824 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909860 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21909836 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153561 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00574912 - -
Whole Mouse Genome G4122A A_51_P140690 AK149129.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P140690 BC057017.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P140690 AB007912.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P140690 AF069708.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P140690 AK002957.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P140690 AK149129.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P140690 BC057017.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P140690 AB007912.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P140690 AF069708.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P140690 AK002957.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123486 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153561 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01497023 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01591330 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01591334 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02096381 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02192371 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02192369 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909836 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909839 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909849 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909788 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909871 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909879 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909813 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909803 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909798 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909840 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909891 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909822 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909749 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909852 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909866 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909801 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909865 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909894 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909772 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909771 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909832 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909884 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909837 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909777 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909833 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909857 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909848 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909888 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909768 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909882 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909826 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909778 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909795 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909854 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909819 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909762 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909815 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909860 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909878 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909869 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909835 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909897 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909814 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909856 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909785 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909823 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909845 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909862 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909769 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909776 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909807 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909783 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909809 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909824 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909791 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909900 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909855 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909886 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909903 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909793 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909797 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909830 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909905 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909765 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909867 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909756 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909828 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909906 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909842 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909786 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909810 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909781 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909804 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21909816 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01497024 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909852 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909807 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909769 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01681649 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909837 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909843 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01465297 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909795 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909778 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909848 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909773 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01886184 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02028019 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909839 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909828 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02028017 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909819 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02185723 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909811 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909776 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909849 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909798 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909791 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909814 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909826 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909834 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909833 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909772 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01629031 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909812 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909797 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909813 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909821 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909815 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909803 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909840 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909809 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909820 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909810 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909847 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909816 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909844 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01497021 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909842 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909771 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02028011 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21909830 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02185716 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01773412 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10490668 - -
Affymetrix Mouse Gene 1.0 ST Array 10490666 - -
Affymetrix Mouse Gene 1.0 ST Array 10490664 - -
Affymetrix Mouse Gene 1.0 ST Array 10490669 - -
Affymetrix Mouse Gene 1.0 ST Array 10490667 - -
Affymetrix Mouse Gene 1.0 ST Array 10490671 - -
Affymetrix Mouse Gene 1.0 ST Array 10490670 - -
Affymetrix HT MG-430 PM Array Plate 1460181_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1435113_PM_x_at - -
Affymetrix Mouse Gene 1.1 ST Array 10490663 - -
Affymetrix Mouse Gene 1.1 ST Array 10490665 - -
Mouse Genome 430 2.0 Array 1435113_x_at - -
Mouse Genome 430 2.0 Array 1435113_PM_x_at - -
Mouse Genome 430 2.0 Array 1460181_at - -
Mouse Genome 430 2.0 Array 1460181_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 20262_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 20262_PM_at - -
Affymetrix Mouse Clariom S Array PSR0200046406.mm.1 NM_009133.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200046406.mm.1 NM_009133.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200046406.mm.1 TC0200005442.mm.1 MTA TC
Mouse Expression Set 430 1435113_PM_x_at - -
Mouse Expression Set 430 1435113_x_at - -
Mouse Expression Set 430 1460181_PM_at - -
Mouse Expression Set 430 1460181_at - -
Murine Genome U74v2 Set 160170_at - -
Murine Genome U74v2 Set 160170_PM_at - -
Murine Genome U74v2 Set 167007_PM_f_at - -
Murine Genome U74v2 Set 165995_PM_r_at - -
Murine Genome U74v2 Set 167007_f_at - -
Murine Genome U74v2 Set 165995_r_at - -
Mouse Genome 430A 2.0 Array 1460181_at - -
Mouse Genome 430A 2.0 Array 1435113_PM_x_at - -
Mouse Genome 430A 2.0 Array 1460181_PM_at - -
Mouse Genome 430A 2.0 Array 1435113_x_at - -
Affymetrix HT Mouse Genome 430A Array 1435113_x_at - -
Affymetrix HT Mouse Genome 430A Array 1460181_at - -
Affymetrix HT Mouse Genome 430A Array 1460181_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1435113_PM_x_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 2690692 - -
Illumina mouse-6 v1.0 expression beadchip 2690692 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002690692 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1225704 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1225704 - -
mouse-6 v1.1 (Illumina) scl020262.2_8-S - -
Illumina Sentrix mouse-6.v1 bead array 2690692 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl020262.2_8-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2690692 - -
MouseRef-8 v2 Expression BeadChip ILMN_1225704 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225704 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl020262.2_8-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0