This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00456117_m1
See other LRIG1 GE Assays ›
Gene Symbol:
Lrig1
Gene Name:
leucine-rich repeats and immunoglobulin-like domains 1
Gene Aliases:
D6Bwg0781e, Img, LIG-1
Chromosome Location:
Chr.6: 94604529 - 94700253 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00456117_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lrig1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008377.2 NP_032403.2 9 - 10 1629 106
GenBank mRNA BC141300.1 - 9 - 10 1421 9055912 106
BC145623.1 - 9 - 10 1421 9054191 106
D78572.1 - 9 - 10 1622 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002773.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lrig1

Panther Classification:

Molecular Function -

immunoglobulin superfamily cell adhesion molecule

Gene Ontology Categories:

Function(s)

sensory perception of sound
otolith morphogenesis
innervation

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600021534.mm.1 NM_008377.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600021534.mm.1 NM_008377.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600021534.mm.1 TC0600002773.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0