This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00455196_g1
See other USP11 GE Assays ›
Gene Symbol:
Usp11
Gene Name:
ubiquitin specific peptidase 11
Gene Aliases:
6230415D12Rik, mKIAA4085
Chromosome Location:
Chr.X: 20703909 - 20720539 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00455196_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Usp11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145628.4 NP_663603.3 5 - 6 710 77
GenBank mRNA AK031767.1 - 5 - 6 689 77
AK049053.1 - 5 - 6 1124 77
AK166523.1 - 5 - 6 686 77
AK220478.1 - 4 - 5 423 77
BC005470.1 - 3 - 4 279 3496607 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00000214.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK031767.1,AK049053.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Usp11

Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
ubiquitin-dependent protein catabolic process
protein deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of proteasomal protein catabolic process
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity
peptidase activity
cysteine-type peptidase activity
hydrolase activity
thiol-dependent ubiquitinyl hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00001441.mm.1 NM_145628.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001441.mm.1 NM_145628.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00001441.mm.1 TC0X00000214.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0