This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00455144_m1
See other NDRG4 GE Assays ›
Gene Symbol:
Ndrg4
Gene Name:
N-myc downstream regulated gene 4
Gene Aliases:
D8Bwg1337e, Ndr1-rs, Ndr4, R74996, SMAP-8
Chromosome Location:
Chr.8: 95676985 - 95715119 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00455144_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndrg4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195006.1 NP_001181935.1 1 - 2 141 80
NM_145602.3 NP_663577.1 1 - 2 141 80
GenBank mRNA AK090374.1 - 1 - 2 123 80
AK144294.1 - 1 - 2 123 80
AK168873.1 - 1 - 2 125 80
BC006595.1 - 1 - 2 115 3156802 80

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK090374.1, AK168873.1, BC006595.1, NM_001195006.1, NM_145602.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndrg4

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
brain development
visual learning
negative regulation of platelet-derived growth factor receptor signaling pathway
positive regulation of neuron projection development
negative regulation of smooth muscle cell migration
vesicle docking
negative regulation of smooth muscle cell proliferation
cell migration involved in heart development
positive regulation of ERK1 and ERK2 cascade
regulation of endocytic recycling
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0