Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300012761.mm.1 |
NM_001146025.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012761.mm.1 |
NM_001146025.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012761.mm.1 |
NM_001146026.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012761.mm.1 |
NM_001146026.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012761.mm.1 |
NM_001146027.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012761.mm.1 |
NM_001146027.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012761.mm.1 |
NM_134064.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012761.mm.1 |
NM_134064.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012761.mm.1 |
NR_027395.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012761.mm.1 |
NR_027395.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012761.mm.1 |
NR_027396.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012761.mm.1 |
NR_027396.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012761.mm.1 |
TC1300002038.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300012760.mm.1 |
NM_001146025.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012760.mm.1 |
NM_001146025.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012760.mm.1 |
NM_001146026.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012760.mm.1 |
NM_001146026.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012760.mm.1 |
NM_001146027.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012760.mm.1 |
NM_001146027.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012760.mm.1 |
NM_134064.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012760.mm.1 |
NM_134064.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012760.mm.1 |
NR_027395.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012760.mm.1 |
NR_027395.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012760.mm.1 |
NR_027396.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012760.mm.1 |
NR_027396.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012760.mm.1 |
TC1300002038.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300012764.mm.1 |
NM_001146025.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012764.mm.1 |
NM_001146025.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012764.mm.1 |
NM_001146026.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012764.mm.1 |
NM_001146026.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012764.mm.1 |
NM_001146027.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012764.mm.1 |
NM_001146027.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012764.mm.1 |
NM_134064.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012764.mm.1 |
NM_134064.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012764.mm.1 |
NR_027395.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012764.mm.1 |
NR_027395.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012764.mm.1 |
NR_027396.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012764.mm.1 |
NR_027396.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300012764.mm.1 |
TC1300002038.mm.1 |
MTA TC |