This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00453821_g1
See other MLC1 GE Assays ›
Gene Symbol:
Mlc1
Gene Name:
megalencephalic leukoencephalopathy with subcortical cysts 1 homolog (human)
Gene Aliases:
AW048630, BB074274, Kiaa0027-hp, LVM, MLC, VL, WKL1, mKIAA0027
Chromosome Location:
Chr.15: 88955884 - 88982687 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00453821_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mlc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133241.2 NP_573504.1 4 - 5 381 69
GenBank mRNA AF449425.1 - 4 - 5 835 69
AK030396.1 - 4 - 5 380 69
AK043757.1 - 4 - 5 381 69
AK122190.1 - 4 - 5 823 69
BC024719.1 - 4 - 5 381 5363238 69

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mlc1

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
protein transport
vesicle-mediated transport
positive regulation of intracellular transport
ion transmembrane transport
regulation of response to osmotic stress
protein oligomerization
cellular response to cholesterol
caveolin-mediated endocytosis
ion channel activity
protein binding
protein transporter activity
protein complex binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0