This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00453160_m1
See other POLH GE Assays ›
Gene Symbol:
Polh
Gene Name:
polymerase (DNA directed), eta (RAD 30 related)
Gene Aliases:
RAD30A, XPV
Chromosome Location:
Chr.17: 46171355 - 46202683 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00453160_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_030715.3 NP_109640.1 1 - 2 231 74
GenBank mRNA AB027128.1 - 1 - 2 257 74
AK036296.1 - 1 - 2 232 74
BC049159.1 - 1 - 2 202 74
BC128366.1 - 1 - 2 228 40111620 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700002167.mm.1

Additional Information:

This assay is in the 5'UTR region of the AB027128.1,AK036296.1,BC128366.1,NM_030715.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polh

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA synthesis involved in DNA repair
DNA replication
DNA repair
pyrimidine dimer repair
postreplication repair
cellular response to DNA damage stimulus
response to UV-C
cellular response to UV-C
DNA biosynthetic process
DNA binding
damaged DNA binding
DNA-directed DNA polymerase activity
transferase activity
nucleotidyltransferase activity
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700020347.mm.1 NM_030715.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700020347.mm.1 NM_030715.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700020347.mm.1 TC1700002167.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700020346.mm.1 NM_030715.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700020346.mm.1 NM_030715.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700020346.mm.1 TC1700002167.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 437491 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0