This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00452294_m1
See other QSOX1 GE Assays ›
Gene Symbol:
Qsox1
Gene Name:
quiescin Q6 sulfhydryl oxidase 1
Gene Aliases:
1300003H02Rik, QSOX, Qscn6, SOx
Chromosome Location:
Chr.1: 155778155 - 155812899 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00452294_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Qsox1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001024945.1 NP_001020116.1 9 - 10 1245 79
NM_023268.2 NP_075757.1 9 - 10 1245 79
GenBank mRNA AB044284.1 - 9 - 10 1153 79
AK004880.1 - 9 - 10 1191 79
AK083938.1 - 9 - 10 1196 79
AK166839.1 - 9 - 10 1191 79
AK169877.1 - 9 - 10 1198 79
AK170449.1 - 9 - 10 1158 79
BC034131.1 - 6 - 7 737 79
BC076590.1 - 9 - 10 1199 30548238 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003309.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Qsox1

Panther Classification:

Molecular Function -

oxidase

Gene Ontology Categories:

Function(s) Process(es)

protein folding
negative regulation of macroautophagy
cell redox homeostasis
oxidation-reduction process
protein disulfide isomerase activity
oxidoreductase activity
flavin-linked sulfhydryl oxidase activity
thiol oxidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100026955.mm.1 NM_001024945.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100026955.mm.1 NM_001024945.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100026955.mm.1 NM_023268.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100026955.mm.1 NM_023268.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100026955.mm.1 TC0100003309.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0