This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00451227_m1
See other TRPC4AP GE Assays ›
Gene Symbol:
Trpc4ap
Gene Name:
transient receptor potential cation channel, subfamily C, member 4 associated protein
Gene Aliases:
4833429F06Rik, D2Ertd113e, Trrp4ap, mFLJ00177, truss
Chromosome Location:
Chr.2: 155634277 - 155692384 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00451227_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trpc4ap
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163452.1 NP_001156924.1 16 - 17 2015 85
NM_019828.2 NP_062802.2 16 - 17 1991 85
GenBank mRNA AF093589.1 - 12 - 13 1487 85
AF130458.1 - 16 - 17 1939 85
AF509881.1 - 16 - 17 1937 85
AK156047.1 - 16 - 17 1964 85
AK170085.1 - 16 - 17 1953 85
AK171404.1 - 16 - 17 1964 85
AK201068.1 - 2 - 3 159 85
AK220239.1 - 16 - 17 1991 85
BC003931.1 - 9 - 10 951 85
BC033274.1 - 16 - 17 1943 3673725 85
BC057330.1 - 16 - 17 1973 6816155 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004967.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trpc4ap

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
protein ubiquitination
phosphatase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200042179.mm.1 NM_001163452.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042179.mm.1 NM_001163452.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042179.mm.1 NM_019828.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042179.mm.1 NM_019828.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200042179.mm.1 TC0200004967.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0