Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100036901.mm.1 |
NM_015810.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036901.mm.1 |
NM_015810.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036901.mm.1 |
NR_027785.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036901.mm.1 |
TC1100003934.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100036898.mm.1 |
NM_015810.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036898.mm.1 |
NM_015810.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036898.mm.1 |
TC1100003934.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100036899.mm.1 |
NM_015810.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036899.mm.1 |
NM_015810.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036899.mm.1 |
NR_027785.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036899.mm.1 |
TC1100003934.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100036895.mm.1 |
NM_015810.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036895.mm.1 |
NM_015810.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036895.mm.1 |
NR_027785.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100036895.mm.1 |
TC1100003934.mm.1 |
MTA TC |