This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00448749_m1
See other SLC34A2 GE Assays ›
Gene Symbol:
Slc34a2
Gene Name:
solute carrier family 34 (sodium phosphate), member 2
Gene Aliases:
AA536683, D5Ertd227e, NaPi-2b, Npt2b
Chromosome Location:
Chr.5: 53049203 - 53071664 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00448749_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc34a2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011402.3 NP_035532.2 1 - 2 42 60
GenBank mRNA AF081499.1 - 1 - 2 39 60
AK004832.1 - 1 - 2 41 60
AK145599.1 - 1 - 2 41 60
AK164786.1 - 1 - 2 43 60
BB873139.1 - 1 - 2 43 60

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000536.mm.1

Additional Information:

This assay is in the 5'UTR region of the AF081499.1,AK004832.1,AK145599.1,NM_011402.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc34a2

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
transport
ion transport
sodium ion transport
phosphate ion transport
cellular phosphate ion homeostasis
phosphate ion transmembrane transport
sodium ion transmembrane transport
sodium-dependent phosphate transport
sodium:phosphate symporter activity
symporter activity
sodium-dependent phosphate transmembrane transporter activity
protein domain specific binding
sodium ion binding
phosphate ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500004970.mm.1 NM_011402.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500004970.mm.1 NM_011402.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500004970.mm.1 TC0500000536.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500004973.mm.1 NM_011402.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500004973.mm.1 NM_011402.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500004973.mm.1 TC0500000536.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0