This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00447484_m1
See other TFAM GE Assays ›
Gene Symbol:
Tfam
Gene Name:
transcription factor A, mitochondrial
Gene Aliases:
AI661103, Hmgts, mtTFA, tsHMG
Chromosome Location:
Chr.10: 71225468 - 71238957 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00447484_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tfam
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009360.4 NP_033386.1 1 - 2 199 130
GenBank mRNA AK004857.1 - 1 - 2 197 130
AK017974.1 - 1 - 2 226 130
AK050446.1 - 1 - 2 200 130
AK161747.1 - 1 - 2 226 130
AK163563.1 - 1 - 2 217 131
AK167348.1 - 1 - 2 200 130
AK167777.1 - 1 - 2 226 130
AK169808.1 - 1 - 2 227 130
BC001987.1 - 1 - 2 194 3487433 130
BC083084.1 - 1 - 2 194 6514962 130
U57939.1 - 1 - 2 191 130
U63859.1 - 1 - 2 399 130

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002356.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tfam

Gene Ontology Categories:

Function(s) Process(es)

chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from mitochondrial promoter
transcription initiation from mitochondrial promoter
mitochondrial respiratory chain complex assembly
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
DNA binding, bending
heat shock protein binding
poly(A) RNA binding
mitochondrial light strand promoter sense binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000016813.mm.1 NM_009360.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000016813.mm.1 NM_009360.4 Refseq
Affymetrix Mouse Clariom S Array PSR1000016813.mm.1 TC1000002356.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000016810.mm.1 NM_009360.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000016810.mm.1 NM_009360.4 Refseq
Affymetrix Mouse Clariom S Array PSR1000016810.mm.1 TC1000002356.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 907099 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0