This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00446388_m1
See other COX4I2 GE Assays ›
Gene Symbol:
Cox4i2
Gene Name:
cytochrome c oxidase subunit IV isoform 2
Gene Aliases:
Cox4b, CoxIV-2
Chromosome Location:
Chr.2: 152754173 - 152765037 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00446388_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cox4i2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053091.2 NP_444321.1 3 - 4 302 61
GenBank mRNA AF271382.1 - 3 - 4 302 61
BC049623.1 - 2 - 3 311 6532529 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002300.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cox4i2

Panther Classification:

Molecular Function -

oxidase

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
hydrogen ion transmembrane transport
cytochrome-c oxidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200017983.mm.1 NM_053091.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017983.mm.1 NM_053091.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200017983.mm.1 TC0200002300.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200017982.mm.1 NM_053091.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200017982.mm.1 NM_053091.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200017982.mm.1 TC0200002300.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0