This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00445306_g1
See other SLC26A3 GE Assays ›
Gene Symbol:
Slc26a3
Gene Name:
solute carrier family 26, member 3
Gene Aliases:
9030623B18Rik, 9130013M11Rik, AV376035, Dra
Chromosome Location:
Chr.12: 31390876 - 31473922 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00445306_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc26a3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021353.3 NP_067328.1 4 - 5 597 67
GenBank mRNA AF136751.1 - 4 - 5 515 67
AK018566.1 - 4 - 5 548 67
AK035716.1 - 2 - 3 550 67
BC022639.1 - 1 - 2 117 67
BC037066.1 - 4 - 5 593 67
BC139271.1 - 3 - 4 432 8862293 67
BC139273.1 - 3 - 4 432 8862295 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200000265.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc26a3

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
chloride transport
sulfate transport
bicarbonate transport
sperm capacitation
intracellular pH elevation
transmembrane transport
membrane hyperpolarization
cellular response to cAMP
anion transmembrane transport
chloride transmembrane transport
protein binding
secondary active sulfate transmembrane transporter activity
bicarbonate transmembrane transporter activity
chloride transmembrane transporter activity
sulfate transmembrane transporter activity
antiporter activity
anion:anion antiporter activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200002086.mm.1 NM_021353.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200002086.mm.1 TC1200000265.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200002080.mm.1 NM_021353.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200002080.mm.1 TC1200000265.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200002081.mm.1 NM_021353.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200002081.mm.1 TC1200000265.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200002085.mm.1 NM_021353.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200002085.mm.1 TC1200000265.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200002087.mm.1 NM_021353.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200002087.mm.1 TC1200000265.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200002082.mm.1 NM_021353.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200002082.mm.1 TC1200000265.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200002079.mm.1 NM_021353.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200002079.mm.1 TC1200000265.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200002084.mm.1 NM_021353.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200002084.mm.1 TC1200000265.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 529150 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0