This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00445277_m1
See other AK3 GE Assays ›
Gene Symbol:
Ak3
Gene Name:
adenylate kinase 3
Gene Aliases:
AA407498, AI506714, AK-3, Ak3l, Ak3l1, Akl3l
Chromosome Location:
Chr.19: 28995380 - 29048672 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00445277_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ak3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021299.1 NP_067274.1 3 - 4 516 75
GenBank mRNA AB020203.1 - 3 - 4 419 75
AK004864.2 - 3 - 4 531 75
AK005194.2 - 3 - 4 570 75
AK007618.1 - 567 75
AK008681.1 - 3 - 4 533 75
AK031631.1 - 3 - 4 514 75
AK053644.1 - 3 - 4 576 75
AK089027.1 - 3 - 4 567 75
AK147060.1 - 3 - 4 576 75
AK149996.1 - 3 - 4 573 75
BC016432.1 - 3 - 4 518 4217820 75
BC019174.1 - 3 - 4 569 5007578 75
BC024871.1 - 3 - 4 516 3154594 75
BC058191.1 - 3 - 4 546 5340253 75

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ak3

Panther Classification:

Molecular Function -

nucleotide kinase

Gene Ontology Categories:

Function(s) Process(es)

nucleobase-containing compound metabolic process
ADP biosynthetic process
AMP phosphorylation
phosphorylation
AMP metabolic process
ATP metabolic process
GTP metabolic process
ITP metabolic process
UTP metabolic process
dATP metabolic process
nucleotide phosphorylation
protein homooligomerization
nucleotide binding
adenylate kinase activity
ATP binding
GTP binding
kinase activity
transferase activity
phosphotransferase activity, phosphate group as acceptor
nucleobase-containing compound kinase activity
identical protein binding
nucleoside triphosphate adenylate kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0