This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00444791_g1
See other PFKP GE Assays ›
Gene Symbol:
Pfkp
Gene Name:
phosphofructokinase, platelet
Gene Aliases:
1200015H23Rik, 9330125N24Rik, ATP-PFK, PFK-C, PFK-P
Chromosome Location:
Chr.13: 6579874 - 6648789 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00444791_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pfkp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291071.1 NP_001278000.1 12 - 13 1307 65
NM_019703.4 NP_062677.1 12 - 13 1307 65
GenBank mRNA AF123533.1 - 12 - 13 1272 65
AK004790.1 - 12 - 13 1261 65
AK049841.1 - 12 - 13 1261 65
AK054193.1 - 12 - 13 1264 65
AK076803.1 - 12 - 13 1262 65
AK098162.1 - 12 - 13 1261 65
AK152670.1 - 12 - 13 1262 65
AK154125.1 - 12 - 13 1262 65
AK165422.1 - 12 - 13 1261 65
AK165425.1 - 12 - 13 1261 65
AK170624.1 - 12 - 13 1262 65
AK171062.1 - 12 - 13 1262 65
AY779275.1 - 12 - 13 1224 65
AY779276.1 - 12 - 13 1224 65
BC006926.1 - 12 - 13 1246 2655383 65
CT010268.1 - 12 - 13 1215 65
Y19008.1 - 12 - 13 1264 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300001395.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pfkp

Panther Classification:

Molecular Function -

carbohydrate kinase kinase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

fructose 6-phosphate metabolic process
glycolytic process
metabolic process
phosphorylation
fructose 1,6-bisphosphate metabolic process
carbohydrate phosphorylation
protein homotetramerization
mitophagy in response to mitochondrial depolarization
nucleotide binding
catalytic activity
6-phosphofructokinase activity
ATP binding
kinase activity
transferase activity
protein complex binding
metal ion binding
fructose-6-phosphate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300008756.mm.1 NM_019703.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300008756.mm.1 TC1300001395.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0