This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00444347_m1
See other ARHGEF7 GE Assays ›
Gene Symbol:
Arhgef7
Gene Name:
Rho guanine nucleotide exchange factor (GEF7)
Gene Aliases:
Cool, PIX, Pak3bp, betaPix, betaPix-b, betaPix-c, cool-1, mKIAA0142, p85Cool1, p85SPR
Chromosome Location:
Chr.8: 11728105 - 11835219 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00444347_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgef7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113517.1 NP_001106989.1 8 - 9 1090 75
NM_001113518.1 NP_001106990.1 7 - 8 879 75
NM_017402.4 NP_059098.2 7 - 8 879 75
GenBank mRNA AF247654.1 - 7 - 8 775 75
AF247655.1 - 5 - 6 496 75
AF343877.1 - 7 - 8 759 75
AK052545.1 - 7 - 8 850 75
AK129064.1 - 8 - 9 1026 75
AK139156.1 - 8 - 9 1019 75
AY220301.1 - 7 - 8 844 75
BC044838.1 - 7 - 8 864 3499604 75
U96634.1 - 5 - 6 569 75

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgef7

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
small GTPase mediated signal transduction
nervous system development
positive regulation of fibroblast migration
lamellipodium assembly
positive regulation of protein binding
regulation of Rho protein signal transduction
intracellular signal transduction
positive regulation of apoptotic process
positive regulation of GTPase activity
astrocyte cell migration
focal adhesion assembly
positive regulation of growth hormone secretion
positive regulation of substrate adhesion-dependent cell spreading
positive regulation of lamellipodium morphogenesis
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
protein kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0