This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00444228_m1
See other CCL20 GE Assays ›
Gene Symbol:
Ccl20
Gene Name:
chemokine (C-C motif) ligand 20
Gene Aliases:
CKb4, LARC, MIP-3A, MIP-3[a], MIP3A, ST38, Scya20, exodus-1
Chromosome Location:
Chr.1: 83116744 - 83119167 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00444228_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ccl20
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016960.2 NP_058656.1 1 - 2 136 89
GenBank mRNA AB015136.1 - 1 - 2 120 89
AF053313.1 - 1 - 2 112 89
AJ222694.1 - 1 - 2 96 89
BX525886.1 - 1 - 2 114 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000718.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Mouse Chemokine Panel Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ccl20

Panther Classification:

Molecular Function -

cytokine intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

chemotaxis
inflammatory response
immune response
positive regulation of interleukin-1 alpha biosynthetic process
positive regulation of nitric-oxide synthase biosynthetic process
cell chemotaxis
positive regulation of T cell migration
cytokine activity
protein binding
chemokine activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100006100.mm.1 NM_001159738.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100006100.mm.1 NM_001159738.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100006100.mm.1 NM_016960.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100006100.mm.1 NM_016960.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100006100.mm.1 TC0100000718.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0