This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00444046_m1
See other DGKA GE Assays ›
Gene Symbol:
Dgka
Gene Name:
diacylglycerol kinase, alpha
Gene Aliases:
80kDa, AW146112, Dagk1
Chromosome Location:
Chr.10: 128720134 - 128748420 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00444046_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dgka
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016811.2 NP_058091.2 15 - 16 1569 72
GenBank mRNA AF085219.1 - 14 - 15 1255 72
AK008272.1 - 4 - 5 241 72
AK008314.1 - 4 - 5 240 72
BC006713.1 - 15 - 16 1569 3584592 72
JN832677.1 - 15 - 16 1512 72
JN832678.1 - 15 - 16 1617 72
JN832679.1 - 15 - 16 1588 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000003161.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dgka

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

protein kinase C-activating G-protein coupled receptor signaling pathway
phosphorylation
intracellular signal transduction
nucleotide binding
diacylglycerol kinase activity
calcium ion binding
ATP binding
phospholipid binding
kinase activity
transferase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000023836.mm.1 NM_016811.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000023836.mm.1 NM_016811.2 Refseq
Affymetrix Mouse Clariom S Array PSR1000023836.mm.1 TC1000003161.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-65c57d65df-v5w74:80/100.66.128.6:80.
git-commit: 123b9e00d0584affee9a3561d252f737ab47bfc1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0