This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00443135_g1
See other SEMA4A GE Assays ›
Gene Symbol:
Sema4a
Gene Name:
sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4A
Gene Aliases:
AI132332, SemB, Semab
Chromosome Location:
Chr.3: 88435959 - 88461182 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00443135_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sema4a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163489.1 NP_001156961.1 5 - 6 523 66
NM_001163490.1 NP_001156962.1 5 - 6 602 66
NM_001163491.1 NP_001156963.1 5 - 6 561 66
NM_013658.3 NP_038686.3 5 - 6 701 66
GenBank mRNA AK077976.1 - 5 - 6 681 66
AK152728.1 - 5 - 6 693 66
AK153164.1 - 5 - 6 693 66
AK154392.1 - 5 - 6 662 66
BC025800.1 - 5 - 6 619 5102015 66
X85991.1 - 5 - 6 523 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002312.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sema4a

Panther Classification:

Molecular Function -

membrane-bound signaling molecule intercellular signal molecule

Gene Ontology Categories:

Function(s)

angiogenesis
T cell differentiation involved in immune response
immune system process
multicellular organism development
nervous system development
axonogenesis
regulation of cell shape
regulation of endothelial cell migration
negative regulation of angiogenesis
cell differentiation
T-helper 1 cell differentiation
semaphorin-plexin signaling pathway

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300017551.mm.1 NM_001163489.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017551.mm.1 NM_001163489.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017551.mm.1 NM_001163490.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017551.mm.1 NM_001163490.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017551.mm.1 NM_001163491.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017551.mm.1 NM_001163491.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017551.mm.1 NM_013658.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300017551.mm.1 NM_013658.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300017551.mm.1 TC0300002312.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0