Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300017559.mm.1 |
NM_001163489.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017559.mm.1 |
NM_001163489.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017559.mm.1 |
NM_001163490.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017559.mm.1 |
NM_001163490.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017559.mm.1 |
NM_001163491.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017559.mm.1 |
NM_001163491.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017559.mm.1 |
NM_013658.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017559.mm.1 |
NM_013658.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017559.mm.1 |
TC0300002312.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300017560.mm.1 |
NM_001163489.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017560.mm.1 |
NM_001163489.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017560.mm.1 |
NM_001163490.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017560.mm.1 |
NM_001163490.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017560.mm.1 |
NM_001163491.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017560.mm.1 |
NM_001163491.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017560.mm.1 |
NM_013658.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017560.mm.1 |
NM_013658.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300017560.mm.1 |
TC0300002312.mm.1 |
MTA TC |