This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00443118_m1
See other SEMA3C GE Assays ›
Gene Symbol:
Sema3c
Gene Name:
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3C
Gene Aliases:
1110036B02Rik, SemE, Semae
Chromosome Location:
Chr.5: 17574816 - 17730268 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00443118_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sema3c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013657.5 NP_038685.3 4 - 5 512 86
GenBank mRNA AK034239.1 - 4 - 5 571 86
AK140348.1 - 1048 86
BC066852.1 - 4 - 5 512 30093230 86
X85994.1 - 4 - 5 464 86

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sema3c

Panther Classification:

Molecular Function -

membrane-bound signaling molecule intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

neural crest cell migration
blood vessel remodeling
multicellular organism development
nervous system development
axon guidance
heart development
post-embryonic development
cell differentiation
dichotomous subdivision of terminal units involved in salivary gland branching
semaphorin receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0